Learn More
Medchemexpress LLC Rosuvastatin (ZD 4522) | 287714-41-4 | MFCD18783208 | 99.7% | 481.54 g/mol | C22H28FN3O6S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17504A10MG
Encompass_Preferred
Rosuvastatin (ZD 4522) is an HMG-CoA reductase inhibitor provided for research and analytical applications. It shows potent biochemical activity and cellular effects and is used as a reference standard in pharmacology and analytical assays. Chemical identifiers: CAS 287714-41-4; formula C22H28FN3O6S; mol wt 481.54 g/mol.
Catalog No. 50-002-98764