Learn More
Grainger Rotary Hammer 11A @ 120V 260 rpm
Supplier:  Grainger 14F135
Encompass_Preferred
The Milwaukee 822AG4 is a 1-9/16" SDS-Max rotary hammer drill that delivers 4.7 ft-lbs of impact energy for heavy-duty drilling and chiseling in concrete and masonry. It has a powerful 12 amp motor that provides 0-1,100 RPM and 0-4,500 BPM. The rotary hammer has a keyless chuck for quick bit changes and an anti-vibration side handle for comfort. It weighs 17.2 lbs and comes with a contractor bag. The 822AG4 is ideal for concrete drilling, chipping and chiseling applications.
Catalog No. 50-274-9186