Learn More
PFEIFFER VACUUM INC Pfeiffer Vacuum Rotary Vane Pump, DuoVane 12 C, 1-phase, Corrosive Duty, Dual-Stage
Supplier:  PFEIFFER VACUUM INC 2000281627
Encompass_Preferred
The DuoVane 12 C Rotary Vane Pump is a robust, two-stage, oil-sealed rotary displacement pump engineered for consistent performance in low and medium vacuum environments. Its advanced design ensures reliable operation, efficiently handling gas compression and expulsion through an oil-sealed exhaust system for superior sealing and lubrication.
  • Reliable and simple to use operation
  • High performance and powerful pumping capabilities
  • Corrosive duty version with a 1-phase motor
  • Includes a gas ballast valve for enhanced performance
  • Two-stage, oil-sealed rotary displacement design
  • Suitable for low and medium vacuum applications
  • Equipped with thermal winding protection for the motor
  • Air (forced convection) cooling method
Catalog No. NC3978946