Learn More
Sigma Aldrich Fine Chemicals Biosciences Rotating 4-way racks Large1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z6759701EA
Encompass
Rota-Rack Tube Racks are fun interlocked racks. Each of the four modules rotates independently. Use several sizes of test tubes culture tubes centrifuge tubes or microcentrifuge tubes at the same time. Click each module into place load with tubes and carry. Rack maintains its orientation until you twist it again. Comes fully assembled. Autoclavable.
Catalog No. 50-290-0890