Learn More
Anton Paar RheolabQC is a rotational rheometer combining high performance with easy operation and robust design. It supports tests from single-point checks to flow curves and yield points, enabling flexible, reliable, and efficient routine rheological measurements
Supplier:  Anton Paar 18318
Encompass_Preferred
SpecificationsSpeed: 0.01 1/min to 1200 1/minTorque: 0.20 mNm to 75 mNmShear stress: 0.5 Pa to 3 x 104 PaShear rate: 10-2 1/s to 6500 1/sViscosity range: (depending on the measuring system)1 mPa.s to 109 mPa.sTemperature range: -20 °C to +180 °CAngular resolution: 2 µradPhysical variables (measurement or evaluation)_Speed_Torque_Shear rate_Shear stress_Dynamic viscosity_Temperature_Time_Deformation_Compliance_Yield stress_ Flow and viscosity curve_Thixotropy_ Time and temperature test
Catalog No. 50-005-80485