Learn More
ATCC Rotavirus A; Strain: WA (TC-adapted)
Supplier:  ATCC VR2018
Encompass
Rotavirus A strain Wa is propagated in MA-104 Clone 1 cells (ATCC CRL-2378.1). This strain was isolated from the diarrhea stool from patient positive for rotavirus and was deposited by National Institute of Allergy and Infectious Disease. It has applications in efficacy testing, virucide testing, and enteric research.
Catalog No. 50-238-4888