Learn More
Medchemexpress LLC Rotenolone | 509-96-6 | 97.0% | 50 MG

Supplier: Medchemexpress LLC HY11969450MG
Rotenolone is an antiproliferative small molecule (CAS 509-96-6) supplied as a powder for research use. It is reported at 97.0% purity, with molecular formula C23H22O7 and molecular weight 410.42 g·mol-1. See product documentation for solubility, storage, and safety data.
- Antiproliferative small molecule suitable for in vitro research.
- CAS 509-96-6 and common synonym 12alpha-hydroxyrotenone.
- Purity 97.0%.
- Molecular formula C23H22O7; molecular weight 410.42 g·mol-1.
- Available as a powder, common vial sizes include 50 MG.
- Soluble in DMSO (100 mg/mL); in vivo formulations documented.
- Store powder at -20°C; store solutions at -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.