Learn More
Apexbio Technology LLC Rotenone, 1g Cas# 83-79-4
SDP
Supplier:  Apexbio Technology LLC B54621000
Encompass_Preferred
inhibitor of the mitochondrial complex I electron transport chain. Rotenone 10mM (in 1mL DMSO). 950455-15-9. MFCD14948691 Rotenone is an inhibitor of the mitochondrial complex I electron transport chain with IC50 value of 1.43283.2 M. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8711