Learn More
Medchemexpress LLC Rotigotine | 99755-59-6 | C19H25NOS | 5 MG
SDP
Supplier:  Medchemexpress LLC HY755025MG
Encompass_Preferred
Rotigotine is a potent dopamine receptor agonist with Ki values of 0.71 nM, 4-15 nM, and 83 nM for the dopamine D3 receptor and D2, D5, D4 receptors and dopamine D1 receptor. It is also a partial agonist of the 5-HT1A receptor and an antagonist of the α2B-adrenergic receptor. Rotigotine can be used for Parkinson's disease (PD) research.
  • Potent dopamine receptor agonist
  • Partial agonist of the 5-HT1A receptor
  • Antagonist of the α2B-adrenergic receptor
  • Useful for Parkinson's disease (PD) research
Catalog No. 50-004-33748