Learn More
Medchemexpress LLC Rotigotine hydrochloride | 125572-93-2 | MFCD00906149 | 99.5% | C19H26ClNOS | 5 MG
SDP
Supplier:  Medchemexpress LLC HYA00075MG
Encompass_Preferred
Rotigotine hydrochloride (N-0923 hydrochloride) is a dopamine receptor full agonist, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor. Intended for research use only, this solid, white to off-white compound has a molecular weight of 351.93 and is soluble in DMSO and H2O.
  • Full agonist of the dopamine receptor.
  • Partial agonist of the 5-HT1A receptor.
  • Antagonist of the α2B-adrenergic receptor.
  • Solid, white to off-white appearance.
  • Soluble in DMSO (≥ 50 mg/mL) and H2O (4.76 mg/mL).
  • Ki of 0.71 nM for the dopamine D3 receptor.
  • Ki of 4-15 nM for the D2, D5, and D4 receptors.
  • Ki of 83 nM for the dopamine D1 receptor.
Catalog No. 50-004-33336