Learn More
Abcam Rotigotine hydrochloride, Dopamine receptor agonist, 50MG
Supplier:  Abcam AB12060150MG
Encompass_Preferred
MW 351.93 g/mol, Purity >99%. Dopamine receptor agonist. High affinity for D₁, D₂ and D₃ receptors (Ki values are 83, 13 and 0.71 nM, respectively). Shows significant affinity at α₂B and 5-HT₁A receptors (Ki values are 27 and 30 nM, respectively). Exhibits anti-Parkinsonian and antidepressant activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7437