Learn More
Cole-Parmer Argos Technologies RotoFlex™ Plus Magnetic Platform; 1/Ea
Supplier:  Cole-Parmer 0439742
Encompass_Preferred
This magnetic platform allows for quick and easy switching of rotisseries for enhanced versatility. Adapt to different racks without tools to accommodate a wide range of tubes. Its compatible with the RotoFlex and RotoFlex Plus systems.
Catalog No. 50-196-0672