Learn More
TARGETMOL CHEMICALS INC Rotundic acid 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T5S050625MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Rotundic acid (Rutundic acid), a natural compound, exhibit cytotoxic activities toward human hepatocellular carcinoma (HepG2), malignant melanoma (A375), SCLC (NCI-H446), breast cancer (MCF-7), and colon cancer (HT-29) cell lines. Purity 98.43%
Catalog No. 50-287-6063