Learn More
MSC Mag-Mate Electromagnet, 1 in Diameter, 24 lb Max Pull, 24 VDC
Supplier:  MSC 01031434
Encompass
This electromagnet features a 1 in diameter and provides a maximum magnetic pull of 24 lb. It operates on 24 VDC and consumes 4.5 W of power. The unit is equipped with a 10-32 thread for easy integration.
  • 1 in Diameter
  • 4.5 W power consumption
  • 24 lb Max magnetic pull
  • 10-32 Thread size
  • 24 VDC operation
Catalog No. 50-005-29869