Learn More
Chemglass Life Sciences 25mL Single Neck Round Bottom Flask (RBF) , 14/20 Outer Joint, #7 Chem-Thread Inlet
SDP
Supplier:  Chemglass Life Sciences CG151420
Encompass_Preferred
25mL Single Neck Round Bottomed Flask, 14/20 Outer Joint, #7 Chem-Thread Inlet. Round bottom, HEAVY WALL flask with a standard taper outer joint and a #7 Chem-Thread as a side neck for insertion of thermometers or bleed tubes having an O.D. between 4 and 7mm.
Catalog No. CG151420