Learn More
Marlin Steel Wire Products Round Mesh Basket with Handles 2.63"D x 2.75"H
SDP
Supplier:  Marlin Steel Wire Products 000036819938
Encompass_Preferred
Rectangular material handling basket with handles for easy gripping.Stacks so that parts inside baskets do not get crushedrobust quality that is ideal for washing parts, degreasing, ultrasonic cleaning, curing, and material handling in a variety of applications such as chemical bath, heat treating, cryogenics, and food processing4 mesh openings per linear inch
Catalog No. NC1959494