Learn More
MSC Vikan Round Hand Scrub, 110 mm, Hard, Red
Supplier:  MSC 47175336
Encompass
This round hand scrub effectively cleans conveyor belts, food containers, and other surfaces. Its ergonomic design allows for increased pressure, making it ideal for demanding tasks in food manufacturing, retail, and restaurant environments.
  • Purpose-built for critical hygiene and food safety in food manufacturing and service, its round design is ideal for cleaning containers.
  • Hard, thick bristles dislodge stubborn soils; ergonomic design with a multi-functional grip reduces worker strain.
  • Durable construction ensures long-lasting performance, easy cleaning, and maintenance.
  • Color-coded for hygienic zoning and features a raised grip to protect hands from chemicals.
  • Autoclavable up to 250 °F (121 °C), dishwasher safe, FDA/USDA compliant, and certified for EU 10/2011 on food contact.
Catalog No. 50-283-9365