Learn More
Grainger Router Bit Set 6 Pc Carbide Tipped
Supplier:  Grainger 16Y554
Encompass_Preferred
The Bosch 15-Piece Router Bit Set includes 15 high speed steel router bits for woodworking applications. The set contains straight, roundover, cove, chamfer, dovetail, and Roman Ogee router bits in 1/4", 3/8", and 1/2" shank diameters. These carbide-tipped bits feature precision ground cutters for clean cuts in wood, plywood, and laminates. The bits come in a plastic carrying case for convenient storage and transport. With its assortment of popular profiles, this router bit set is ideal for woodworkers looking for a starter set of bits for edge treatments and joinery.
Catalog No. 50-276-7851