Learn More
Grainger Router Corded 1.25 hp
Supplier:  Grainger 14F181
Encompass_Preferred
The Grainger Approved 1.25 HP Corded Router is a powerful and versatile woodworking tool. It has a 1.25 HP motor that provides up to 27,000 RPM for smooth, accurate cuts. The fixed base has a depth adjustment ring that allows for plunge cuts up to 1-3/4" deep. It accepts 1/4" and 1/2" shank router bits and has a quick release cam lock system for easy bit changes. The soft start motor and variable speed control dial provide enhanced performance and control. This router also features a dust sealed switch that prevents dust ingestion to extend switch life. It comes with a 1 year warranty.
Catalog No. 50-275-0113