Learn More
Essendant ROUTERMESH WIFIWH
Supplier:  Essendant LNKWHW0302
Encompass_Preferred
Modular design performs well within every home type situation and footprint Each node is a powerful Tri-Band connection point offering up to 2 000 square feet of coverage Simply add additional nodes to work around walls problem areas or multiple stories Designed for high-performance Wi-Fi Each one features a beautiful sleek design that can be placed in open spaces and away from obstructions Compact in size it is easy to place throughout the home and features a smaller footprint than that of a traditional router Its tower design supports vertical antenna placement which enhances Wi-Fi dispersion throughout your home Additionally it also features a cable management system designed to hide connected cables and keep your home clutter-free Pack of Two Network Types- 802 11ac Wireless Maximum Wireless Range- 200 ft Number of Ports- 1 Connects Devices (From/To)- Router to Wireless Network
Catalog No. 50-000-32795