Learn More
Cayman Chemical RoxatIdIn Acetatehydrochlo 1g
SDP
Supplier:  Cayman Chemical 212481 G
Encompass_Preferred
A competitive histamine H2 receptor antagonist upon conversion to its active metabolite reduces both intracellular cAMP concentrations and gastric acid secretion by parietal cells
Catalog No. 50-298-1438