Learn More
Medchemexpress LLC Roxindole hydrochloride | 108050-82-4 | MFCD09265259 | 99.0% | 382.93 g/mol | C23H27ClN2O | 10 MG

Supplier: Medchemexpress LLC HY106100A10MG
Roxindole hydrochloride is the hydrochloride salt of roxindole, a dopaminergic and serotonergic research compound used in receptor pharmacology and neuropharmacology studies. It functions primarily as a dopamine autoreceptor agonist with activity at multiple dopamine receptor subtypes and serotonergic sites, and is provided as solid material or as a DMSO solution for laboratory use.
- Dopamine autoreceptor agonist with activity at D2-like receptors.
- Shows affinity for 5-HT1A serotonergic sites.
- Supplied as solid and solution pack sizes for experimental flexibility.
- Molecular weight 382.93 g/mol; formula C23H27ClN2O.
- Storage recommended at -20°C; protect from light and store under inert gas.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.