Learn More
Medchemexpress LLC HY-139609 5mg , RP-3500 CAS:2417489-10-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1396095MG
Encompass_Preferred
HY-139609 5mg RP-3500 CAS: 2417489-10-0 RP-3500 (ATR inhibitor 4) is an orally active, selective ATR kinase inhibitor (ATRi) with an IC50 of 1.00 nM in biochemical assays. RP-3500 shows 30-fold selectivity for ATR over mTOR (IC50=120 nM) and >2,000-fold selectivity over ATM, DNA-PK, and PI3Kα kinases. RP-3500 has potent antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9145