Learn More
Medchemexpress LLC Rp 70676 | 136609-26-2 | MFCD00907619 | 99.7% | 416.58 | C25H28N4S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10157625MG
Encompass_Preferred
RP 70676 is a small-molecule inhibitor of acyl-coenzyme A:cholesterol acyltransferase (ACAT) supplied for laboratory research (CAS 136609-26-2). It is characterized in enzyme and cell assays and is available as solid material or as ready-to-use DMSO solutions for dosing and assay development.
  • Potent ACAT inhibition in vitro (rat IC50 25 nM; rabbit IC50 44 nM).
  • Active across species and tissue assays including hamster liver and human hepatic tissue.
  • Shows cell-based activity in macrophage assays (P388D1 IC50 ~540 nM; J774 inhibition 0.09 μM).
  • Provided as solid and pre-made DMSO solutions to simplify dosing and handling.
  • High reported purity (~99.7%).
  • For research use only; not for human or clinical applications.
Catalog No. 50-002-48510