Learn More
Medchemexpress LLC (Rp,R'p)-(R)-Mandyphos | 210842-74-3 | 92.6% | C52H50FeN2P2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY79503500MG
Encompass_Preferred
(Rp,R'p)-(R)-Mandyphos is a chemical compound with the formula C52H50FeN2P2 and a molecular weight of 820.76. It is a drug intermediate primarily used for the synthesis of various active compounds and has a purity of 92.59%. This product is intended for research use only.
  • Solid appearance, light yellow to yellow color
  • Store at 4°C, protected from light and under nitrogen
  • For in solvent storage, maintain at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen
  • Shipped at room temperature
Catalog No. 50-004-33014