Learn More
Cayman Chemical RpAdenosIn5O 1thIotrIp 2.6mg
SDP
Supplier:  Cayman Chemical 385352.6 MG
Encompass_Preferred
An isomer of ATP-α-S and a P2Y1 receptor agonist; increases calcium mobilization in HEK293 cells expressing the human P2Y1 receptor (EC50 = 75 nM); binds to washed isolated human platelets (Ki = 156 nM); inhibits ADP-induced aggregation of, and PGE1-induced cAMP production in, isolated human PRP (pA2s = 4.74 and 5.26, respectively); induces relaxation of carbamoylcholine-preconstricted guinea pig taenia coli strips (EC50 = 56 nM); has been used in the synthesis of cyclic dinucleotides that are recognized by bacterial riboswitches
Catalog No. 50-291-9001