Learn More
Medchemexpress LLC RPI-1 | 269730-03-2 | MFCD03852474 | 99.3% | 297.31 g·mol⁻¹ | C17H15NO4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101246100MG
Encompass_Preferred
RPI-1 is a 2-indolinone small-molecule inhibitor of the RET tyrosine kinase used for laboratory research. It inhibits RET autophosphorylation and downstream signaling (PLCγ, ERK, AKT) in cellular models and has demonstrated antitumor activity. Supplied as a solid or as a solution for use in biochemical and cell-based assays.
  • Selective RET tyrosine kinase inhibition in cellular assays.
  • Reduces RET phosphorylation and downstream signaling pathways.
  • High reported purity suitable for biochemical studies.
  • Available in multiple sizes and formats for experimental flexibility.
  • Provided with certificate of analysis and safety data documentation.
Catalog No. 50-002-11112