Learn More
Cole-Parmer Benchtop Tubing Cutter, 1/16" to 5/8" OD
Supplier:  Cole-Parmer 5011848
Encompass_Preferred
Use tubing cutters to make clean, straight cuts through your plastic tubing. Straight cuts ensure better connection to fittings.
Catalog No. 50-255-7740