Learn More
Meso Scale Discovery RPLEX HUM CD30/TNFRSF8 ASYQP
SDP
Supplier:  Meso Scale Discovery K151M5R22
Encompass
The R-PLEX Human CD30 (TNFRSF8) Assay is a multiplex immunoassay panel that measures CD30 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and is compatible with the SECTOR Imager 6000 The assay has a detection range of 0 03 - 400 ng/mL and the minimum recommended sample dilution is 2-fold It includes a plate coated with capture antibodies detection antibodies labeled with ECL tags and calibrators and controls The assay allows high-throughput sensitive and reproducible quantification of CD30 a cell membrane protein belonging to the tumor necrosis factor receptor family
Catalog No. 50-000-79955