Learn More
Meso Scale Discovery RPLEX HUM IGF2R/CIM6PR ASYQP
SDP
Supplier:  Meso Scale Discovery K151C4R22
Encompass
The R-PLEX Human IGF-2R (CI-MPR) Assay is a multiplex immunoassay kit that enables simultaneous measurement of Insulin-like Growth Factor 2 Receptor (IGF-2R) concentrations in human serum plasma and cell culture supernatants This assay utilizes Meso Scale Discovery electrochemiluminescence technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent and calibrators and controls This assay allows efficient and accurate quantification of IGF-2R levels in research studies
Catalog No. 50-000-82470