Learn More
Meso Scale Discovery RPLEX HUM Influenza B NP ASYQP
SDP
Supplier:  Meso Scale Discovery K1519HR22
Encompass
The R-PLEX Human Influenza B NP Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that simultaneously detects influenza B nucleoprotein antigens in human nasopharyngeal swab specimens This assay utilizes electrochemiluminescence detection technology and provides quantitative results for influenza B in under 3 hours It is intended for research use only and not for use in diagnostic procedures The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags assay diluent wash buffer read buffer and calibrators and controls to allow quantitative analysis of samples
Catalog No. 50-000-80328