Learn More
Meso Scale Discovery RPLEX HUM Neurofilament ASYQP
SDP
Supplier:  Meso Scale Discovery K1517XR22
Encompass
The Meso Scale Discovery R-Plex Human Neurofilament L Assay is a multiplex immunoassay that quantitatively measures neurofilament light chain (NfL) in human serum plasma and CSF This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 10 000-fold greater than ELISA The assay requires 50 L or less sample volume has a 3 5 hour total assay time and shows no cross-reactivity with other biomarkers The R-Plex Human Neurofilament L Assay delivers reliable reproducible results for the quantification of NfL an important biomarker of axonal injury and neurodegeneration This assay is ideal for neurology research and clinical trials
Catalog No. 50-000-80550