Learn More
Meso Scale Discovery RPLEX HUM Procalcitonin ASYQP
SDP
Supplier:  Meso Scale Discovery K151AJTR22
Encompass
The R-PLEX Human Procalcitonin Assay is a multiplex immunoassay panel that measures Procalcitonin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and is compatible with the SECTOR S 600 instrument The assay has a reportable range of 0 04 - 75 ng/mL and the total assay time is approximately 2 hours This assay provides a precise accurate and sensitive measurement of Procalcitonin which can aid in the diagnosis and monitoring of severe systemic bacterial fungal or parasitic infections The R-PLEX assay requires a low sample volume of 50 L and has high lot-to-lot reproducibility
Catalog No. 50-000-81662