Learn More
Meso Scale Discovery RPLEX HUM Serpi A12Vaspi ASYQP
SDP
Supplier:  Meso Scale Discovery K1510SR22
Encompass
The R-PLEX Human Serpin A12 (Vaspin) Assay Kit from Meso Scale Discovery is an immunoassay that quantitatively measures human Serpin A12 (Vaspin) in serum plasma and cell culture supernatants This assay kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The total assay time is just under 3 5 hours This product can be used to study Serpin A12 (Vaspin) biology and its role as an adipokine
Catalog No. 50-000-78384