Learn More
Meso Scale Discovery RPLEX MSE Osteoproteger ASYQP
SDP
Supplier:  Meso Scale Discovery K152U3R22
Encompass
The Meso Scale Discovery R-PLEX Mouse Osteoprotegerin Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of mouse osteoprotegerin (OPG) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on Meso Scale Discovery SECTOR Imagers The assay can be used to study bone metabolism osteoporosis and effects of various treatments on OPG levels The R-PLEX assay provides reproducible and quantitatively precise data compared to ELISA assays
Catalog No. 50-000-78665