Learn More
Bioworld RPMI-1640 Medium w/L-glutamine and 25 mM HEPES, 1 L
Supplier:  Bioworld 306111692
Encompass_Preferred
DescriptionRPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy's 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells.Specification:LiquidWith L-Glutamine and 25mM HEPESCell Culture tested
Catalog No. 50-254-244
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.