Learn More
ATCC RPMI 2650; Nasal Septum Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CCL30
Encompass
RPMI 2650 is a cell line exhibiting epithelial morphology from the nasal septum of a 52-year-old, male patient with squamous cell carcinoma. This cell line was deposited by GE Moore and can be used in cancer research, and toxicology.
Catalog No. 50-238-4068