Learn More
ATCC RPMI 6666; Hodgkins Lymphoma; Human (Homo sapiens)
Supplier:  ATCC CCL113
Encompass
RPMI 6666 is a cell line exhibiting lymphoblast morphology that was isolated from the peripheral blood of a White, 29-year-old, male patient with Hodgkins disease. This cell line was deposited by GE Moore and can be used in immunology research.
Catalog No. 50-238-3240