Learn More
Medchemexpress LLC RPR132595A hydrochloride | 185304-42-1 | MFCD18252420 | 99.5% | 555.02 g/mol | C28H31ClN4O6 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY100638A1MG
Encompass_Preferred
RPR132595A hydrochloride is the hydrochloride salt of RPR132595A (NPC), an active metabolite of irinotecan (CPT-11) formed by cytochrome P-450 3A4 (CYP3A4) and primarily excreted in urine. It is supplied as a research reagent and reference standard for metabolic, pharmacokinetic, and analytical studies.
  • Active metabolite of irinotecan produced by cytochrome P-450 3A4.
  • Hydrochloride salt form for improved stability and handling.
  • High purity suitable for use as a reference standard in analytical assays.
  • Soluble in DMSO at experimental concentrations.
  • Available in research-scale quantities for laboratory studies.
Catalog No. 50-002-13855