Learn More
ABclonal Technology RPS27A Rabbit pAb
SDP
Supplier:  ABclonal Technology A146181000UL
Encompass_Preferred
This gene encodes a fusion protein consisting of ubiquitin at the N terminus and ribosomal protein S27a at the C terminus. Ubiquitin plays a key role in targeting proteins for degradation by the 26S proteasome. When expressed in yeast, the fusion protein is processed into free ubiquitin and ribosomal protein S27a. S27a is part of the 40S ribosomal subunit and belongs to the S27AE family of ribosomal proteins. It contains C4-type zinc finger domains and is located in the cytoplasm. Pseudogenes of this gene are found in the genome. Similar fusion proteins, such as ribosomal protein L40 with ubiquitin and S30 with fubi, are also synthesized. Multiple alternatively spliced transcript variants encoding the same proteins have been identified.
Catalog No. 50-001-07105