Learn More
Cayman Chemical RpThymIdIn5O 1thIotrIp 2.5mg
SDP
Supplier:  Cayman Chemical 386892.5 MG
Encompass_Preferred
An isomer of TTP-α-S; binds to HIV-1 reverse transcriptase (Kds = 45.7, 27.32, and 39.44 µM in the presence of magnesium, manganate, and cobalt, respectively)
Catalog No. 50-291-8988