Learn More
Medchemexpress LLC HY-W035409 5mg Medchemexpress, RPW-24 CAS:1001625-82-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0354095MG
Encompass_Preferred
Medchemexpress, HY-W035409 5mg RPW-24 CAS:1001625-82-6 RPW-24 protects C. elegans from bacterial infection by stimulating the host immune response of the nematode. RPW-24 has antibacterial activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0739