Learn More
TARGETMOL CHEMICALS INC RQ-00203078 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T200725MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. RQ-00203078 is a highly selective, potent, and orally available TRPM8 antagonist.
Catalog No. 50-287-7457