Learn More
Medchemexpress LLC Rr-11a analog | 685543-66-2 | 99.1% | 491.49 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY112205A
Encompass_Preferred
RR-11a analog is a potent and irreversible inhibitor of Schistosoma mansoni legumain, with an IC50 of 31 nM. It is an aza-Asn derivative and an aza-peptide Michael acceptor.
  • Potent and irreversible inhibitor of Schistosoma mansoni legumain
  • IC50 of 31 nM against Schistosoma mansoni legumain
  • Aza-Asn derivative
  • Aza-peptide Michael acceptor
  • Available for research use only
Catalog No. 50-004-38812