Learn More
Strem, An Ascensus Company CAS 880262-16-8. 2g. ((4R5R)-()-O-[1-Benzyl-1-(5-methyl-2-phenyl-45-dihydrooxazol-4-yl)-2-phenylethyl] (diphenylphosphinite)(15-COD)iridium(I) tetrakis(35-bis(trifluoromethyl)phenylborate min. 97% (RR)-[COD]Ir[Ph2PThrePHOX]. MFCD18253385
SDP
Supplier:  Strem, An Ascensus Company 7750192G
Encompass
CAS 880262-16-8. 2g. ((4R5R)-()-O-[1-Benzyl-1-(5-methyl-2-phenyl-45-dihydrooxazol-4-yl)-2-phenylethyl] (diphenylphosphinite)(15-COD)iridium(I) tetrakis(35-bis(trifluoromethyl)phenylborate min. 97% (RR)-[COD]Ir[Ph2PThrePHOX]. MFCD18253385. Molecular Weight 1719.25. Molecular Formula Ir(C8H12)(C37H34NO2P)B[C6H3(CF3)2]4ae. Color/form orange pwdr. Strem 77-5019. http//www.strem.com/catalog/v/77-5019/
Catalog No. 50-211-0853