Learn More
Carbosynth LLC. H-Arg-Arg-Leu-Ile-Glu-Asp-Ala-Glu-Tyr-Ala-Ala-Arg-Gly-NH2, 5mg, MW: 1519.69, Molecular formula: C64H106N22O21, CAS# 81156-93-6
SDP
Supplier:  Carbosynth LLC. PTK3956PI5MG
Encompass_Preferred
Synonyms: RR-SCR, RRLIEDAEYAARG, Application: Substrate for Protein Tyrosine Kinase (PTK), Salt Form:, Acetate Form, Storage Temperature: -20°C, References: J.E. Casnellie, M.L. Harrison, L.J. Pike, K.E. Hellstrom, and E.G. Krebs, Proc. Natl. Acad. Sci. USA 79, 282 (1982)., S.E. Ramer, D.G. Winker, A. Carrera, T.M. Robets, and C.T. Walsh, Proc. Natl. Acad. Sci. USA, 88, 6254, (1991)., H.S. Earp, K.S. Austin, G.Y. Gillespie, S.C. Buessow, A.a. Davies, and P.J. Parker, J. Biol. Chem., 260, 4351 (1985).
Catalog No. 50-168-6795