Learn More
Selleck Chemical LLC RRx-001 5mg 925206-65-1
SDP
Supplier:  Selleck Chemical LLC S84055MG
Encompass_Preferred
RRx-001 is a novel epigenetic modulator with potential radiosensitizing activity. It inhibits glucose 6-phosphate dehydrogenase(G6PD) in human tumor cells, binds hemoglobin and drives RBC-mediated redox reactions under hypoxia. RRx-001 triggers apoptosis and exhibits anticancer activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6036