Learn More
Apexbio Technology LLC RS 09 1449566-36-2 25mg
SDP
Supplier:  Apexbio Technology LLC B879725
Encompass_Preferred
RS 09 (CAS 1449566-36-2) is a synthetic peptide that functions as a lipopolysaccharide (LPS) mimetic It modulates immune signaling pathways by acting as an agonistic adjuvant resulting in altered biological responses Experimental studies have demonstrated that RS 09 can enhance vaccine-induced antibody production in in vitro systems Due to its defined immunomodulatory activity RS 09 is widely employed in immunology research and vaccine development to investigate mechanisms of immune activation and to optimize adjuvant formulations
Catalog No. 50-000-89384