Learn More
Medchemexpress LLC HY-P1439 5mg , RS 09 CAS:1449566-36-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP14395MG
Encompass_Preferred
HY-P1439 5mg RS 09 CAS: 1449566-36-2 RS09 is a LPS peptide mimic serves as a candidate to be considered as a new class of TLR4 agonist adjuvant. RS09 increases antibody production in a vaccine setting. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1571