Learn More
Cayman Chemical RS 09trIfluoroacetAT salt 10mg
SDP
Supplier:  Cayman Chemical 3315210 MG
Encompass_Preferred
A TLR4 agonist; activates NF-κB in a concentration-dependent manner in a SEAP assay using TLR4-expressing HEK293 cells and induces NF-κB nuclear translocation in the same cells; acts as an adjuvant in mice vaccinated with a KLH-conjugated X-15 antigen at 25 UG/animal
Catalog No. 50-294-7343